Gene Transcripts

Gene Symbol
OR14C36
Entrez Gene ID
127066
Gene Name
olfactory receptor family 14 subfamily C member 36
Gene Aliases
OR5BF1
Chromosome Location
Chr. 1: 248348775 - 248349713 on Build GRCh38
UniGene ID
Hs.553583
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001918.1 NP_001001918.1 1 213

View Full Product Details