Gene Transcripts
- Gene Symbol
- SLC41A1
- Entrez Gene ID
- 254428
- Gene Name
- solute carrier family 41 member 1
- Gene Aliases
- MgtE
- Chromosome Location
- Chr. 1: 205789093 - 205813759 on Build GRCh38
- UniGene ID
- Hs.20274
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173854.5 |
NP_776253.3
|
8, 9 |
2102 |
|
XM_005245069.2 |
XP_005245126.1
|
8, 9 |
1778 |
|
GenBank mRNA |
AJ514402.1 |
- |
7, 8 |
1189 |
|
AK125075.1 |
- |
7, 8 |
863 |
|
AK128069.1 |
- |
7, 8 |
1355 |
|
AK314732.1 |
- |
7, 8 |
1260 |
|
AL831974.1 |
- |
8, 9 |
1938 |
|
AL832001.1 |
- |
8, 9 |
1938 |
|
AL832362.1 |
- |
8, 9 |
1938 |
|
BC136303.1 |
- |
7, 8 |
1583 |
9020290 |
BX641054.1 |
- |
8, 9 |
1940 |
|
BX648979.1 |
- |
8, 9 |
1976 |
|
View Full Product Details