Gene Transcripts

Gene Symbol
PCSK9
Entrez Gene ID
255738
Gene Name
proprotein convertase subtilisin/kexin type 9
Gene Aliases
FH3, HCHOLA3, LDLCQ1, NARC-1, NARC1, PC9
Chromosome Location
Chr. 1: 55039476 - 55064853 on Build GRCh38
UniGene ID
Hs.18844
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174936.3 NP_777596.2 4 966
NR_110451.1 2 263
GenBank mRNA AK075365.1 - 4 895
AK122717.1 - 5 651
AK124635.1 - 1 1425
AK293870.1 - 5 946
AK297473.1 - 2 263
EF692496.1 - 4 604

View Full Product Details