Gene Transcripts

Gene Symbol
UNC80
Entrez Gene ID
285175
Gene Name
unc-80 homolog, NALCN activator
Gene Aliases
C2orf21, UNC-80
Chromosome Location
Chr. 2: 209771925 - 209999300 on Build GRCh38
UniGene ID
Hs.396201
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032504.1 NP_115893.1 5, 6 798
NM_182587.3 NP_872393.3 5, 6 798
XM_005246476.1 XP_005246533.1 5, 6 798
XM_011511004.1 XP_011509306.1 5, 6 798
XM_011511005.1 XP_011509307.1 5, 6 798
XM_011511006.1 XP_011509308.1 5, 6 798
XM_011511007.1 XP_011509309.1 5, 6 798
XM_011511008.1 XP_011509310.1 5, 6 798
XM_011511010.2 XP_011509312.1 5, 6 798
XM_011511012.1 XP_011509314.1 5, 6 798
XM_017003884.1 XP_016859373.1 5, 6 798
XM_017003885.1 XP_016859374.1 5, 6 798
XM_017003886.1 XP_016859375.1 5, 6 798
XM_017003887.1 XP_016859376.1 5, 6 798
XM_017003888.1 XP_016859377.1 5, 6 798
XM_017003889.1 XP_016859378.1 5, 6 798
XM_017003890.1 XP_016859379.1 5, 6 798
XM_017003891.1 XP_016859380.1 4, 5 817
XM_017003893.1 XP_016859382.1 5, 6 798
GenBank mRNA AK090815.1 - 6, 7 798
BC136690.1 - 5, 6 769
BC136693.1 - 5, 6 769
DW009748.1 - 1, 2 121

View Full Product Details