Gene Transcripts

Gene Symbol
LINC00636
Entrez Gene ID
285205
Gene Name
long intergenic non-protein coding RNA 636
Gene Aliases
Chromosome Location
Chr. 3: 107883205 - 107928907 on Build GRCh38
UniGene ID
Hs.134882
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015394.1 3 764
GenBank mRNA BC039517.1 - 3 763
BC101228.2 - 3 722
BC101229.1 - 3 721
BC101230.2 - 3 721
BC101231.1 - 3 720

View Full Product Details