Gene Transcripts

Gene Symbol
OTOG
Entrez Gene ID
340990
Gene Name
otogelin
Gene Aliases
DFNB18B, MLEMP, OTGN
Chromosome Location
Chr. 11: 17547373 - 17645944 on Build GRCh38
UniGene ID
Hs.688380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277269.1 NP_001264198.1 29 3576
NM_001292063.1 NP_001278992.1 30 3540
GenBank mRNA AK128214.1 - 7 905

View Full Product Details