Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001018056.2 | NP_001018066.1 | 8 | 1577 | |
NM_001322225.1 | NP_001309154.1 | 7 | 1454 | ||
NM_001322226.1 | NP_001309155.1 | 7 | 1454 | ||
NM_003383.4 | NP_003374.3 | 8 | 1577 | ||
GenBank mRNA | AB208822.1 | - | 9 | 1333 | |
AK092381.1 | - | 6 | 844 | ||
AK095744.1 | - | 4 | 657 | ||
BC136562.1 | - | 8 | 1298 | 9020552 | |
BC142653.1 | - | 8 | 1239 | 40148037 | |
BC144245.1 | - | 8 | 1298 | 9052765 | |
D16493.1 | - | 8 | 1231 | ||
D16494.1 | - | 8 | 1162 | ||
L20470.1 | - | 8 | 1552 | ||
L22431.1 | - | 8 | 1274 | ||
S73849.1 | - | 8 | 1246 |