Gene Transcripts

Gene Symbol
GP1BA
Entrez Gene ID
2811
Gene Name
glycoprotein Ib platelet alpha subunit
Gene Aliases
BDPLT1, BDPLT3, BSS, CD42B, CD42b-alpha, DBPLT3, GP1B, GPIbA, GPIbalpha, VWDP
Chromosome Location
Chr. 17: 4932275 - 4935030 on Build GRCh38
UniGene ID
Hs.1472
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000173.6 NP_000164.5 2 1028
GenBank mRNA BC027955.1 - 2 1006
J02940.1 - 2 973
KC120774.1 - 2 1006
KC120775.1 - 2 1006
KC120776.1 - 2 1005

View Full Product Details