Gene Transcripts
- Gene Symbol
- FUS
- Entrez Gene ID
- 2521
- Gene Name
- FUS RNA binding protein
- Gene Aliases
- ALS6, ETM4, FUS1, HNRNPP2, POMP75, TLS
- Chromosome Location
- Chr. 16: 31179444 - 31194871 on Build GRCh38
- UniGene ID
- Hs.46894
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001170634.1 |
NP_001164105.1
|
9, 10 |
1031 |
|
NM_001170937.1 |
NP_001164408.1
|
9, 10 |
1022 |
|
NM_004960.3 |
NP_004951.1
|
9, 10 |
1034 |
|
NR_028388.2 |
|
8, 9 |
999 |
|
XM_005255233.4 |
XP_005255290.1
|
8, 9 |
1662 |
|
XM_011545781.1 |
XP_011544083.1
|
9, 10 |
1000 |
|
XM_011545782.2 |
XP_011544084.1
|
4, 5 |
390 |
|
GenBank mRNA |
AB208902.1 |
- |
7, 8 |
4104 |
|
AK098774.1 |
- |
9, 10 |
1002 |
|
AK130774.1 |
- |
8, 9 |
971 |
|
AK290936.1 |
- |
9, 10 |
1002 |
|
AK299129.1 |
- |
9, 10 |
790 |
|
BC000402.2 |
- |
9, 10 |
997 |
2822692 |
BC002459.1 |
- |
9, 10 |
969 |
3345294 |
BC026062.2 |
- |
9, 10 |
987 |
3937437 |
BT007131.1 |
- |
8, 9 |
929 |
|
CR456747.1 |
- |
8, 9 |
929 |
|
S62140.1 |
- |
9, 10 |
1007 |
|
U36561.1 |
- |
8, 9 |
938 |
|
X71428.1 |
- |
9, 10 |
998 |
|
View Full Product Details