Gene Transcripts

Gene Symbol
ASXL1
Entrez Gene ID
171023
Gene Name
additional sex combs like 1, transcriptional regulator
Gene Aliases
BOPS, MDS
Chromosome Location
Chr. 20: 32358062 - 32439319 on Build GRCh38
UniGene ID
Hs.374043
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015338.5 NP_056153.2 11 1447
XM_006723727.3 XP_006723790.1 10 1726
XM_006723728.3 XP_006723791.1 11 1056
XM_006723730.3 XP_006723793.1 11 1252
XM_006723732.2 XP_006723795.1 10 1075
XM_006723733.1 XP_006723796.1 4 1081
XM_011528648.2 XP_011526950.1 11 1371
XM_011528652.2 XP_011526954.1 12 2389
XM_017027704.1 XP_016883193.1 11 2512
XM_017027705.1 XP_016883194.1 11 1525
XM_017027706.1 XP_016883195.1 10 1576
GenBank mRNA AB023195.2 - 5 497
AB084280.1 - 10 1000
AJ438952.2 - 11 1273
AK122923.1 - 10 1075
AL117518.1 - 4 417
BC137278.1 - 12 1210 9021275
BC137280.1 - 12 1210 9021277

View Full Product Details