Gene Transcripts

Gene Symbol
GAS2
Entrez Gene ID
2620
Gene Name
growth arrest specific 2
Gene Aliases
Chromosome Location
Chr. 11: 22626113 - 22885930 on Build GRCh38
UniGene ID
Hs.711297
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143830.1 NP_001137302.1 2 440
NM_005256.3 NP_005247.1 1 231
NM_177553.2 NP_808221.1 2 382
XM_011519971.2 XP_011518273.1 2 440
XM_011519972.2 XP_011518274.1 2 2435
XM_011519975.1 XP_011518277.1 2 440
XM_011519978.2 XP_011518280.1 2 440
XM_017017528.1 XP_016873017.1 2 1132
XM_017017529.1 XP_016873018.1 2 277
XM_017017530.1 XP_016873019.1 2 470
XM_017017531.1 XP_016873020.1 2 291
XM_017017532.1 XP_016873021.1 2 440
GenBank mRNA AK313734.1 - 2 446
AU099094.1 - 1 231
BC013326.1 - 2 386
BC040470.1 - 2 382 4280621
CR450285.1 - 1 134
DA345287.1 - 2 440
U95032.1 - 1 200

View Full Product Details