Gene Transcripts
- Gene Symbol
- LOC105378397
- Entrez Gene ID
- 105378397
- Gene Name
- uncharacterized LOC105378397
- Gene Aliases
- Chromosome Location
- Chr. 10: 83672406 - 83677089 on Build GRCh38
- UniGene ID
- Hs.147725
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_134317.1 |
|
3, 4 |
247 |
|
GenBank mRNA |
BX092644.1 |
- |
3, 4 |
247 |
|
View Full Product Details