Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013495.2 | NP_038523.2 | 14 | 1758 | |
GenBank mRNA | AF017175.1 | - | 1646 | ||
AK077454.1 | - | 14 | 1712 | ||
AK128925.1 | - | 14 | 1712 | ||
AK133472.1 | - | 14 | 1713 | ||
AK136487.1 | - | 14 | 1712 | ||
AK147770.1 | - | 14 | 1713 | ||
AK149688.1 | - | 14 | 1709 | ||
BC038395.1 | - | 13 | 1693 | 2649352 | |
BC046383.1 | - | 13 | 1693 | 4160676 | |
BC054791.1 | - | 14 | 1739 | 6832731 |