Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253392.1 | NP_001240321.1 | 19 | 2516 | |
NM_007958.1 | NP_031984.1 | 21 | 2977 | ||
GenBank mRNA | AK052406.1 | - | 1 | 1240 | |
AK122454.1 | - | 21 | 2784 | ||
BC042442.1 | - | 19 | 2509 | 3980726 | |
CF531941.1 | - | 3 | 260 | ||
X69942.1 | - | 21 | 2977 |