Gene Transcripts
- Gene Symbol
- Psme3
- Entrez Gene ID
- 19192
- Gene Name
- proteaseome (prosome, macropain) activator subunit 3 (PA28 gamma, Ki)
- Gene Aliases
- AA410043, AU020960, Ki, PA28gamma, REGgamma, pa28g
- Chromosome Location
- Chr. 11: 101316251 - 101323531 on Build GRCm38
- UniGene ID
- Mm.288477
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011192.3 |
NP_035322.1
|
8, 9 |
735 |
|
GenBank mRNA |
AK140597.1 |
- |
8, 9 |
756 |
|
AK150939.1 |
- |
8, 9 |
755 |
|
AK156079.1 |
- |
8, 9 |
705 |
|
AK171348.1 |
- |
8, 9 |
751 |
|
BC015255.1 |
- |
8, 9 |
735 |
4221225 |
BC139818.1 |
- |
8, 9 |
683 |
40141268 |
CT010198.1 |
- |
8, 9 |
539 |
|
D87911.1 |
- |
8, 9 |
658 |
|
U60330.1 |
- |
8, 9 |
539 |
|
View Full Product Details