Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000576.2 | NP_000567.1 | 5 | 440 | |
XM_017003988.1 | XP_016859477.1 | 4 | 441 | ||
GenBank mRNA | AK313983.1 | - | 4 | 1160 | |
BC008678.1 | - | 5 | 440 | 3875593 | |
BT007213.1 | - | 4 | 353 | ||
CR407679.1 | - | 4 | 353 | ||
K02770.1 | - | 5 | 439 | ||
M15330.1 | - | 5 | 439 | ||
M54933.1 | - | 5 | 440 | ||
X02532.1 | - | 5 | 430 | ||
X56087.1 | - | 5 | 410 |