Gene Transcripts

Gene Symbol
Dmrt1
Entrez Gene ID
50796
Gene Name
doublesex and mab-3 related transcription factor 1
Gene Aliases
Chromosome Location
Chr. 19: 25505618 - 25604328 on Build GRCm38
UniGene ID
Mm.391208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015826.5 NP_056641.2 3 923
GenBank mRNA AB221653.1 - 3 775
AF202778.1 - 3 910
AL133300.1 - 3 926
AY169786.1 - 3 926
AY169787.1 - 3 926
AY943925.1 - 3 465
BC125474.1 - 3 886 40129989

View Full Product Details