Gene Transcripts

Gene Symbol
4930453N24Rik
Entrez Gene ID
67609
Gene Name
RIKEN cDNA 4930453N24 gene
Gene Aliases
AI649104
Chromosome Location
Chr. 16: 64762152 - 64771156 on Build GRCm38
UniGene ID
Mm.212157
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026273.2 NP_080549.1 3 970
GenBank mRNA AK075826.1 - 3 954
AK161311.1 - 3 971
BC020029.1 - 3 959 4012943

View Full Product Details