Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198834.2 | NP_942131.1 | 20 | 3020 | |
NM_198836.2 | NP_942133.1 | 20 | 2588 | ||
NM_198837.1 | NP_942134.1 | 18 | 2658 | ||
NM_198838.1 | NP_942135.1 | 19 | 2769 | ||
NM_198839.2 | NP_942136.1 | 24 | 3694 | ||
XM_005257267.4 | XP_005257324.1 | 20 | 2501 | ||
XM_006721853.1 | XP_006721916.1 | 20 | 3031 | ||
XM_011524701.1 | XP_011523003.1 | 20 | 2941 | ||
XM_011524703.1 | XP_011523005.1 | 20 | 2573 | ||
XM_011524704.2 | XP_011523006.1 | 20 | 2506 | ||
XM_017024553.1 | XP_016880042.1 | 21 | 3055 | ||
XM_017024554.1 | XP_016880043.1 | 21 | 3055 | ||
XM_017024555.1 | XP_016880044.1 | 20 | 2654 | ||
GenBank mRNA | AB371587.1 | - | 20 | 2567 | |
AK309084.1 | - | 20 | 2576 | ||
AY237919.1 | - | 18 | 2397 | ||
AY315627.1 | - | 18 | 2397 | ||
BC137287.1 | - | 20 | 2525 | 9021284 | |
U19822.1 | - | 18 | 2397 |