Gene Transcripts

Gene Symbol
2210011C24Rik
Entrez Gene ID
70134
Gene Name
RIKEN cDNA 2210011C24 gene
Gene Aliases
Chromosome Location
Chr. 8: 84010228 - 84011720 on Build GRCm38
UniGene ID
Mm.41017
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291292.1 NP_001278221.1 3 863
GenBank mRNA AI847924.1 - 2 492
AK008705.1 - 3 582

View Full Product Details