Gene Transcripts

Gene Symbol
Atad2
Entrez Gene ID
70472
Gene Name
ATPase family, AAA domain containing 2
Gene Aliases
2610509G12Rik
Chromosome Location
Chr. 15: 58094047 - 58135139 on Build GRCm38
UniGene ID
Mm.221758
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027435.2 NP_081711.2 25 3558
GenBank mRNA AK029867.1 - 18 2705
BC032211.1 - 1 92
BC071261.1 - 2 122

View Full Product Details