Gene Transcripts

Gene Symbol
Lrrc69
Entrez Gene ID
73314
Gene Name
leucine rich repeat containing 69
Gene Aliases
1700034K16Rik
Chromosome Location
Chr. 4: 14623622 - 14801320 on Build GRCm38
UniGene ID
Mm.52613
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028499.2 NP_082775.1 4, 5 594
GenBank mRNA AK006601.1 - 4, 5 605
AK161371.1 - 4, 5 597

View Full Product Details