Gene Transcripts

Gene Symbol
Zdbf2
Entrez Gene ID
73884
Gene Name
zinc finger, DBF-type containing 2
Gene Aliases
4930431J08Rik, 9330107J05Rik
Chromosome Location
Chr. 1: 63200358 - 63314576 on Build GRCm38
UniGene ID
Mm.86986
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267872.1 NP_001254801.1 6 1120
NM_001285936.1 NP_001272865.1 7 1254
NM_001285937.1 NP_001272866.1 6 1197
NM_028673.1 NP_082949.1 7 1170
GenBank mRNA AB777269.1 - 6 1197
AB777270.1 - 7 1254
AK132808.1 - 6 1121

View Full Product Details