Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133753.1 | NP_598514.1 | 4 | 1471 | |
GenBank mRNA | AK004851.1 | - | 4 | 1490 | |
AK139780.1 | - | 4 | 1488 | ||
AK166791.1 | - | 4 | 1490 | ||
AK167156.1 | - | 4 | 1430 | ||
AK167359.1 | - | 4 | 1490 | ||
BC005546.1 | - | 4 | 1471 | 3585640 | |
BC057646.1 | - | 4 | 1489 | 4194124 |