Gene Transcripts

Gene Symbol
Kbtbd13
Entrez Gene ID
74492
Gene Name
kelch repeat and BTB (POZ) domain containing 13
Gene Aliases
5430433E21Rik
Chromosome Location
Chr. 9: 65388684 - 65391652 on Build GRCm38
UniGene ID
Mm.159223
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028974.1 NP_083250.1 1 1058
GenBank mRNA AK017385.1 - 1 71
AK048611.1 - 1 951
BC139793.1 - 2 938

View Full Product Details