Gene Transcripts

Gene Symbol
4930451I11Rik
Entrez Gene ID
78118
Gene Name
RIKEN cDNA 4930451I11 gene
Gene Aliases
Chromosome Location
Chr. 7: 126830468 - 126831540 on Build GRCm38
UniGene ID
Mm.107753
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183131.2 NP_898954.2 2 209
GenBank mRNA AK019625.1 - 2 186
BC048622.1 - 2 209 6743640

View Full Product Details