Gene Transcripts
- Gene Symbol
- C1qtnf3
- Entrez Gene ID
- 81799
- Gene Name
- C1q and tumor necrosis factor related protein 3
- Gene Aliases
- 2310005P21Rik, CORS-26, CORS26, CTRP3, Corcs, Cors
- Chromosome Location
- Chr. 15: 10952332 - 10980162 on Build GRCm38
- UniGene ID
- Mm.280158
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001204134.1 |
NP_001191063.1
|
5, 6 |
899 |
|
NM_030888.4 |
NP_112150.1
|
5, 6 |
705 |
|
GenBank mRNA |
AF246265.1 |
- |
5, 6 |
682 |
|
AK029352.1 |
- |
5, 6 |
682 |
|
AK075867.1 |
- |
5, 6 |
682 |
|
AK081704.1 |
- |
5, 6 |
682 |
|
AK128898.1 |
- |
5, 6 |
705 |
|
BC019636.1 |
- |
5, 6 |
908 |
|
BC055047.1 |
- |
5, 6 |
667 |
6486507 |
BC055859.1 |
- |
5, 6 |
646 |
4238617 |
DQ002396.1 |
- |
5, 6 |
583 |
|
View Full Product Details