Gene Transcripts

Gene Symbol
Ano6
Entrez Gene ID
105722
Gene Name
anoctamin 6
Gene Aliases
2900059G15Rik, AA407480, AW554778, F730003B03Rik, Tmem16f
Chromosome Location
Chr. 15: 95790843 - 95975472 on Build GRCm38
UniGene ID
Mm.38087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253813.1 NP_001240742.1 5 510
NM_175344.4 NP_780553.2 4 447
GenBank mRNA AK089300.1 - 4 358
AK159411.1 - 5 461
BC060732.1 - 4 447 6409332

View Full Product Details