Gene Transcripts

Gene Symbol
KCNJ10
Entrez Gene ID
3766
Gene Name
potassium voltage-gated channel subfamily J member 10
Gene Aliases
BIRK-10, KCNJ13-PEN, KIR1.2, KIR4.1, SESAME
Chromosome Location
Chr. 1: 160037467 - 160070261 on Build GRCh38
UniGene ID
Hs.408960
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002241.4 NP_002232.2 2 2396
GenBank mRNA BC034036.1 - 2 2298
BC131627.1 - 2 2292 40118533

View Full Product Details