Gene Transcripts

Gene Symbol
SLC29A4
Entrez Gene ID
222962
Gene Name
solute carrier family 29 member 4
Gene Aliases
ENT4, PMAT
Chromosome Location
Chr. 7: 5274311 - 5304073 on Build GRCh38
UniGene ID
Hs.4302
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040661.1 NP_001035751.1 11 2573
NM_001300847.1 NP_001287776.1 11 2528
NM_153247.2 NP_694979.2 11 2570
XM_005249658.4 XP_005249715.1 11 2494
XM_006715667.3 XP_006715730.1 11 2441
XM_006715668.2 XP_006715731.1 11 2519
XM_011515200.2 XP_011513502.1 11 2601
XM_011515201.2 XP_011513503.1 11 2590
GenBank mRNA AK075422.1 - 9 2147
AK092242.1 - 11 2556
BC025325.1 - 11 2429 3354432
BC047592.1 - 11 2533 4816109

View Full Product Details