Gene Transcripts
- Gene Symbol
- RHOA
- Entrez Gene ID
- 387
- Gene Name
- ras homolog family member A
- Gene Aliases
- ARH12, ARHA, RHO12, RHOH12
- Chromosome Location
- Chr. 3: 49359136 - 49412097 on Build GRCh38
- UniGene ID
- Hs.247077
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001313941.1 |
NP_001300870.1
|
4, 5 |
646 |
|
NM_001313943.1 |
NP_001300872.1
|
3, 4 |
551 |
|
NM_001313944.1 |
NP_001300873.1
|
3, 4 |
491 |
|
NM_001313945.1 |
NP_001300874.1
|
3, 4 |
428 |
|
NM_001664.3 |
NP_001655.1
|
3, 4 |
551 |
|
GenBank mRNA |
AF498970.1 |
- |
2, 3 |
271 |
|
AK130066.1 |
- |
3, 4 |
547 |
|
AK130808.1 |
- |
3, 4 |
427 |
|
AK222556.1 |
- |
3, 4 |
443 |
|
AK296654.1 |
- |
3, 4 |
367 |
|
AK307220.1 |
- |
3, 4 |
326 |
|
BC001360.2 |
- |
3, 4 |
383 |
3051460 |
BC005976.1 |
- |
3, 4 |
388 |
4102976 |
BF966305.1 |
- |
4, 5 |
521 |
|
BQ721697.1 |
- |
3, 4 |
424 |
|
BT019870.1 |
- |
2, 3 |
271 |
|
BX647063.1 |
- |
3, 4 |
447 |
|
L09159.1 |
- |
2, 3 |
260 |
|
L25080.1 |
- |
3, 4 |
422 |
|
X05026.1 |
- |
3, 4 |
429 |
|
View Full Product Details