Gene Transcripts

Gene Symbol
RHOA
Entrez Gene ID
387
Gene Name
ras homolog family member A
Gene Aliases
ARH12, ARHA, RHO12, RHOH12
Chromosome Location
Chr. 3: 49359136 - 49412097 on Build GRCh38
UniGene ID
Hs.247077
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001313941.1 NP_001300870.1 6 924
NM_001313943.1 NP_001300872.1 6 968
NM_001313944.1 NP_001300873.1 5 769
NM_001313945.1 NP_001300874.1 5 706
NM_001313946.1 NP_001300875.1 3 577
NM_001313947.1 NP_001300876.1 4 708
NM_001664.3 NP_001655.1 5 829
GenBank mRNA AF498970.1 - 4 549
AK130066.1 - 5 825
AK130808.1 - 5 705
AK222556.1 - 5 721
AK296654.1 - 5 645
AK307220.1 - 5 604
BC000946.3 - 2 273
BC001360.2 - 5 661 3051460
BC005976.1 - 5 666 4102976
BT019870.1 - 4 549
BU509012.1 - 4 539
BX647063.1 - 5 725
L09159.1 - 4 538
L25080.1 - 5 700
LN607834.1 - 2 275
X05026.1 - 5 707

View Full Product Details