Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282161.1 | NP_001269090.1 | 6 | 1296 | |
NM_003336.3 | NP_003327.2 | 6 | 1303 | ||
NM_181762.2 | NP_861427.1 | 5 | 1213 | ||
GenBank mRNA | AK223045.1 | - | 6 | 1205 | |
AK223064.1 | - | 6 | 1202 | ||
AK297696.1 | - | 5 | 1115 | ||
AK301675.1 | - | 5 | 1214 | ||
BC042021.1 | - | 3 | 2080 | ||
M74524.1 | - | 6 | 1178 |