Gene Transcripts

Gene Symbol
DND1
Entrez Gene ID
373863
Gene Name
DND microRNA-mediated repression inhibitor 1
Gene Aliases
RBMS4
Chromosome Location
Chr. 5: 140670796 - 140673586 on Build GRCh38
UniGene ID
Hs.591262
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194249.2 NP_919225.1 4 1418
GenBank mRNA BC033496.1 - 4 1418

View Full Product Details