Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025194.2 | NP_079470.1 | 4 | 1565 | |
XM_017027324.1 | XP_016882813.1 | 5 | 749 | ||
GenBank mRNA | AJ290975.1 | - | 4 | 1532 | |
AK024596.1 | - | 4 | 600 | ||
BC060788.1 | - | 4 | 1565 | 30332196 | |
D38169.1 | - | 4 | 1296 |