Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278656.1 | NP_001265585.1 | 2 | 499 | |
NM_004175.4 | NP_004166.1 | 2 | 513 | ||
NR_103819.1 | 2 | 513 | |||
GenBank mRNA | AB451249.1 | - | 1 | 73 | |
AB451373.1 | - | 1 | 73 | ||
AK296173.1 | - | 2 | 156 | ||
BC000457.2 | - | 2 | 169 | 2822042 | |
BC003150.1 | - | 2 | 169 | 2822042 | |
BC034447.1 | - | 2 | 196 | ||
BF344644.1 | - | 2 | 508 | ||
BM928709.1 | - | 2 | 461 | ||
BP202359.1 | - | 2 | 163 | ||
BY993733.1 | - | 2 | 185 | ||
CN286602.1 | - | 2 | 153 | ||
CR456583.1 | - | 2 | 189 | ||
U15009.1 | - | 2 | 160 |