Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011274.5 | NP_035404.4 | 8 | 1316 | |
GenBank mRNA | AF091096.1 | - | 6 | 838 | |
AK052034.1 | - | 3 | 497 | ||
AK133383.1 | - | 8 | 991 | ||
AK155994.1 | - | 8 | 1290 | ||
AK162285.1 | - | 3 | 490 | ||
AK165045.1 | - | 1 | 268 | ||
AK166566.1 | - | 8 | 1307 | ||
BC023029.1 | - | 8 | 976 | 5374731 |