Gene Transcripts

Gene Symbol
Slc29a4
Entrez Gene ID
243328
Gene Name
solute carrier family 29 (nucleoside transporters), member 4
Gene Aliases
ENT4, mPMAT
Chromosome Location
Chr. 5: 142692476 - 142722490 on Build GRCm38
UniGene ID
Mm.125942
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146257.2 NP_666369.1 4 515
GenBank mRNA BC025599.1 - 4 459
BY727407.1 - 4 516

View Full Product Details