Gene Transcripts

Gene Symbol
HIST1H1D
Entrez Gene ID
3007
Gene Name
histone cluster 1, H1d
Gene Aliases
H1.3, H1D, H1F3, H1s-2
Chromosome Location
Chr. 6: 26234212 - 26234988 on Build GRCh38
UniGene ID
Hs.136857
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005320.2 NP_005311.1 1 690
GenBank mRNA AK312845.1 - 1 689
BC104874.1 - 1 716 8143877
BC111971.1 - 1 716 8327439

View Full Product Details