Gene Transcripts

Gene Symbol
Dolpp1
Entrez Gene ID
57170
Gene Name
dolichyl pyrophosphate phosphatase 1
Gene Aliases
0610011H20Rik, AB030189, LSFR2
Chromosome Location
Chr. 2: 30392124 - 30400529 on Build GRCm38
UniGene ID
Mm.285167
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290508.1 NP_001277437.1 3, 4 566
NM_001290509.1 NP_001277438.1 2, 3 359
NM_020329.4 NP_065062.1 2, 3 359
GenBank mRNA AB030189.1 - 2, 3 199
AK002544.1 - 3, 4 405
AK044141.1 - 2, 3 198
AK053923.1 - 2, 3 188
AK088065.1 - 2, 3 203
BB853200.1 - 2, 3 362
BC026544.1 - 2, 3 183 4975127
BY133699.1 - 2, 3 284

View Full Product Details