Gene Transcripts

Gene Symbol
Ces2g
Entrez Gene ID
72361
Gene Name
carboxylesterase 2G
Gene Aliases
2210023G05Rik, Ces2l
Chromosome Location
Chr. 8: 104961611 - 104969537 on Build GRCm38
UniGene ID
Mm.26580
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_197999.2 NP_932116.2 12 2299
GenBank mRNA BC026641.1 - 4 1055
BC027185.1 - 12 2277 4194872
BC027555.1 - 1 94

View Full Product Details