Gene Transcripts
- Gene Symbol
- Sorbs2
- Entrez Gene ID
- 114901
- Gene Name
- sorbin and SH3 domain containing 2
- Gene Aliases
- Argbp2
- Chromosome Location
- Chr. 16: 49626205 - 49820223 on Build Rnor_6.0
- UniGene ID
- Rn.24612
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053770.1 |
NP_446222.1
|
8, 9 |
1244 |
|
GenBank mRNA |
AF026505.1 |
- |
8, 9 |
1244 |
|
View Full Product Details