Gene Transcripts

Gene Symbol
Rnf40
Entrez Gene ID
266712
Gene Name
ring finger protein 40, E3 ubiquitin protein ligase
Gene Aliases
Chromosome Location
Chr. 1: 199037481 - 199052823 on Build Rnor_6.0
UniGene ID
Rn.162466
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153471.1 NP_703201.1 3, 4 400
XM_006230219.2 XP_006230281.1 3, 4 463
GenBank mRNA AF352815.1 - 3, 4 400

View Full Product Details