Gene Transcripts

Gene Symbol
Cxcr1
Entrez Gene ID
54258
Gene Name
chemokine (C-X-C motif) receptor 1
Gene Aliases
Il8ra
Chromosome Location
Chr. 9: 81466443 - 81469299 on Build Rnor_6.0
UniGene ID
Rn.138115
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019310.1 NP_062183.1 1 551
XM_006245273.2 XP_006245335.2 2 628

View Full Product Details