Gene Transcripts
- Gene Symbol
- Slc12a1
- Entrez Gene ID
- 25065
- Gene Name
- solute carrier family 12 (sodium/potassium/chloride transporter), member 1
- Gene Aliases
- Nkcc2
- Chromosome Location
- Chr. 3: 117421530 - 117498372 on Build Rnor_6.0
- UniGene ID
- Rn.14799
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270617.1 |
NP_001257546.1
|
2, 3 |
622 |
|
NM_001270618.1 |
NP_001257547.1
|
2, 3 |
622 |
|
XM_008762140.1 |
XP_008760362.1
|
2, 3 |
694 |
|
XM_008762141.1 |
XP_008760363.1
|
2, 3 |
693 |
|
XM_008762142.1 |
XP_008760364.1
|
2, 3 |
694 |
|
XM_008762143.1 |
XP_008760365.1
|
2, 3 |
694 |
|
XM_008762144.1 |
XP_008760366.1
|
2, 3 |
694 |
|
GenBank mRNA |
BC087017.1 |
- |
2, 3 |
664 |
|
EF577032.1 |
- |
1, 2 |
606 |
|
FM059245.1 |
- |
1, 2 |
254 |
|
JN579708.1 |
- |
2, 3 |
622 |
|
U10096.1 |
- |
2, 3 |
622 |
|
View Full Product Details