Gene Transcripts
- Gene Symbol
- NARF
- Entrez Gene ID
- 26502
- Gene Name
- nuclear prelamin A recognition factor
- Gene Aliases
- IOP2
- Chromosome Location
- Chr. 17: 82458184 - 82490537 on Build GRCh38
- UniGene ID
- Hs.256526
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001083608.1 |
NP_001077077.1
|
1, 2 |
168 |
|
NM_012336.3 |
NP_036468.1
|
1, 2 |
168 |
|
NM_031968.2 |
NP_114174.1
|
1, 2 |
168 |
|
XM_006722278.3 |
XP_006722341.1
|
1, 2 |
256 |
|
XM_011523543.2 |
XP_011521845.1
|
1, 2 |
74 |
|
XM_011523544.2 |
XP_011521846.1
|
1, 2 |
265 |
|
GenBank mRNA |
AF128406.1 |
- |
1, 2 |
119 |
|
AK000929.1 |
- |
1, 2 |
121 |
|
AK297875.1 |
- |
1, 2 |
168 |
|
AK303155.1 |
- |
1, 2 |
121 |
|
AK307351.1 |
- |
1, 2 |
121 |
|
BC000438.2 |
- |
1, 2 |
135 |
2820621 |
BC016440.1 |
- |
1, 2 |
55 |
3447710 |
DB123829.1 |
- |
1, 2 |
195 |
|
View Full Product Details