Gene Transcripts
- Gene Symbol
- WDR62
- Entrez Gene ID
- 284403
- Gene Name
- WD repeat domain 62
- Gene Aliases
- C19orf14, MCPH2
- Chromosome Location
- Chr. 19: 36054881 - 36111145 on Build GRCh38
- UniGene ID
- Hs.116244
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001083961.1 |
NP_001077430.1
|
17, 18 |
2241 |
|
NM_173636.4 |
NP_775907.4
|
17, 18 |
2241 |
|
XM_005258809.2 |
XP_005258866.1
|
17, 18 |
2241 |
|
XM_011526837.1 |
XP_011525139.1
|
17, 18 |
2226 |
|
XM_011526838.1 |
XP_011525140.1
|
17, 18 |
2241 |
|
XM_011526839.1 |
XP_011525141.1
|
15, 16 |
1890 |
|
XM_011526840.2 |
XP_011525142.1
|
10, 11 |
1236 |
|
XM_011526841.2 |
XP_011525143.1
|
8, 9 |
857 |
|
XM_011526842.1 |
XP_011525144.1
|
7, 8 |
689 |
|
XM_017026665.1 |
XP_016882154.1
|
17, 18 |
2241 |
|
GenBank mRNA |
BC058939.1 |
- |
17, 18 |
2185 |
|
BQ880028.1 |
- |
4, 5 |
410 |
|
BX647726.1 |
- |
18, 19 |
2302 |
|
View Full Product Details