Gene Transcripts

Gene Symbol
Sorbs2
Entrez Gene ID
114901
Gene Name
sorbin and SH3 domain containing 2
Gene Aliases
Argbp2
Chromosome Location
Chr. 16: 49626205 - 49820223 on Build Rnor_6.0
UniGene ID
Rn.24612
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053770.1 NP_446222.1 8 1165
GenBank mRNA AF026505.1 - 8 1165

View Full Product Details