Gene Transcripts

Gene Symbol
Wipf1
Entrez Gene ID
117538
Gene Name
WAS/WASL interacting protein family, member 1
Gene Aliases
Waspip, Wip
Chromosome Location
Chr. 3: 60150001 - 60207125 on Build Rnor_6.0
UniGene ID
Rn.161803
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057192.2 NP_476540.2 2 302
GenBank mRNA AJ303456.1 - 1 135
BC072475.1 - 2 302 7100751

View Full Product Details