gRNA Design Details
- Target DNA Sequence
- CGGCGTGTACCTCTTCACAG
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 19132431 - 19132453 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.12/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TMEM161A
- Entrez Gene ID
-
54929
- Gene Name
- transmembrane protein 161A
- Gene Aliases
-
AROS-29
- Chromosome Location
-
Chr.19: 19119616 - 19138501 on GRCh38
- UniGene ID
- Hs.631629
- Species
-
Human